Få mere at vide
N-Acetyl-S-propyl-L-cysteine, TRC
Artikelnummer. 30179851
Skift visning
Mængde:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30179851 10 mg
30179784 50 mg
30179840 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30179851 Leverandør Toronto Research Chemicals Leverandørnr. TRCA16803510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 14402-54-1
Molekylær formel C8 H15 N O3 S
Molekylvægt (g/mol) 205.27
Synonym L-Cysteine, N-acetyl-S-propyl- (9CI, ACI), Alanine, N-acetyl-3-(propylthio)- (6CI, 7CI), Alanine, N-acetyl-3-(propylthio)-, L- (8CI), N-Acetyl-S-propyl-L-cysteine (ACI), (R)-2-Acetamido-3-(propylthio)propanoic acid, Propylmercapturic acid, (2R)-2-Acetamido-3-(propylsulfanyl)propanoic acid
IUPAC navn (2R)-2-acetamido-3-propylsulfanylpropanoic acid
SMIL CCCSC[C@H](NC(=O)C)C(=O)O
Tekniske data
Kemisk navn eller materiale N-Acetyl-S-propyl-L-cysteine
Renhedsgrad noter HPLC
InChI formel InChI=1S/C8H15NO3S/c1-3-4-13-5-7(8(11)12)9-6(2)10/h7H,3-5H2,1-2H3,(H,9,10)(H,11,12)/t7-/m0/s1
Mængde 10 mg
Formel vægt 205.0773
Procent renhed >95%