Få mere at vide
N-Acetyl-S-(2-cyanoethyl)-L-cysteine Ammonium Salt, TRC
Artikelnummer. 30009391
Skift visning
Mængde:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30009391 25 mg
17937204 5 mg
17949484 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30009391 Leverandør Toronto Research Chemicals Leverandørnr. TRCA17208325MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 168208-30-8
Molekylær formel C8H11N2O3S.H4N
Molekylvægt (g/mol) 233.29
Synonym N-Acetyl-S-(2-cyanoethyl)cysteine Ammonium Salt, S-(2-Cyanoethyl)mercapturic Acid Ammonium Salt;
IUPAC navn ammonium;(2R)-2-acetamido-3-(2-cyanoethylsulfanyl)propanoate
SMIL [NH4+].CC(=O)N[C@@H](CSCCC#N)C(=O)[O-]
Tekniske data
Kemisk navn eller materiale N-Acetyl-S-(2-cyanoethyl)-L-cysteine Ammonium Salt
Renhedsgrad noter HPLC
InChI formel InChI=1S/C8H12N2O3S.H3N/c1-6(11)10-7(8(12)13)5-14-4-2-3-9;/h7H,2,4-5H2,1H3,(H,10,11)(H,12,13);1H3/t7-;/m0./s1
Mængde 25 mg
Formel vægt 233.0834
Procent renhed >95%