Få mere at vide
N-Acetyl-S-(1-cyano-2-hydroxyethyl)-L-cysteine Dicyclohexylamine Salt (Mixture of Diastereomers), TRC
Artikelnummer. 17993795
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17993795 10 mg
17972115 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17993795 Leverandør Toronto Research Chemicals Leverandørnr. TRCA17208010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1356930-89-6
Molekylær formel C12 H23 N . C8 H12 N2 O4 S
Molekylvægt (g/mol) 413.57
Synonym (2R)-2-Acetamido-3-(1-cyano-2-hydroxyethyl)sulfanylpropanoic acid, N-cyclohexylcyclohexanamine, N-Acetyl-S-(1-cyano-2-hydroxyethyl)-L-cysteine dicyclohexylamine salt, N-Acetyl-S-(1-cyano-2-hydroxyethyl)cysteine dicyclohexylamine salt
IUPAC navn (2R)-2-acetamido-3-(1-cyano-2-hydroxy-ethyl)sulfanyl-propanoic acid;N-cyclohexylcyclohexanamine
SMIL CC(=O)N[C@@H](CSC(CO)C#N)C(=O)O.C1CCC(CC1)NC2CCCCC2
Tekniske data
Kemisk navn eller materiale N-Acetyl-S-(1-cyano-2-hydroxyethyl)-L-cysteine Dicyclohexylamine Salt (Mixture of Diastereomers)
InChI formel InChI=1S/C12H23N.C8H12N2O4S/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12;1-5(12)10-7(8(13)14)4-15-6(2-9)3-11/h11-13H,1-10H2;6-7,11H,3-4H2,1H3,(H,10,12)(H,13,14)/t;6?,7-/m.0/s1
Mængde 10 mg
Formel vægt 413.2348