Få mere at vide
N-Acetyl Demethyl Memantine, TRC
Artikelnummer. 17960535
Skift visning
Mængde:
25 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17960535 50 mg
17902875 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17960535 Leverandør Toronto Research Chemicals Leverandørnr. TRCA17351050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 778-09-6
Molekylær formel C13 H21 N O
Molekylvægt (g/mol) 207.3119
Synonym N-(3-Methyltricyclo[3.3.1.13,7]dec-1-yl)acetamide, N-(3-Methyl-1-adamantyl)acetamide, 1-Acetamido-3-methyladamantane, 1-Methyl-3-acetamidoadamantane, NSC 179381
IUPAC navn N-[(5R,7S)-3-methyl-1-adamantyl]acetamide
SMIL CC(=O)N[C@@]12C[C@H]3C[C@H](C[C@@](C)(C3)C1)C2
Tekniske data
Kemisk navn eller materiale N-Acetyl Demethyl Memantine
Mængde 50 mg
InChI formel InChI=1S/C13H21NO/c1-9(15)14-13-6-10-3-11(7-13)5-12(2,4-10)8-13/h10-11H,3-8H2,1-2H3,(H,14,15)/t10-,11+,12-,13-
Formel vægt 207.1623