Få mere at vide
N-Acetyl-D-allo-isoleucine, TRC
Artikelnummer. 30179286
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30179286 10 mg
30178988 100 mg
30178979 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30179286 Leverandør Toronto Research Chemicals Leverandørnr. TRCA16819310MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 54831-20-8
Molekylær formel C8H15NO3
Molekylvægt (g/mol) 173.21
IUPAC navn (2R,3S)-2-acetamido-3-methylpentanoic acid
SMIL CC[C@H](C)[C@@H](NC(=O)C)C(=O)O
Tekniske data
Kemisk navn eller materiale N-Acetyl-D-allo-isoleucine
InChI formel InChI=1S/C8H15NO3/c1-4-5(2)7(8(11)12)9-6(3)10/h5,7H,4H2,1-3H3,(H,9,10)(H,11,12)/t5-,7+/m0/s1
Mængde 10 mg
Formel vægt 173.1052