Få mere at vide
N-Acetyl Axitinib, TRC
Artikelnummer. 30137762
Skift visning
Mængde:
2.5 mg
10 mg
25 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30137762 2.5 mg 2.5mg
30135198 10 mg -
30011314 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30137762 Leverandør Toronto Research Chemicals Leverandørnr. TRCA1345602.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1639137-80-6
Molekylær formel C24H20N4O2S
Molekylvægt (g/mol) 428.51
Synonym (E)-2-((1-Acetyl-3-(2-(pyridin-2-yl)vinyl)-1H-indazol-6-yl)thio)-N-methylbenzamide
IUPAC navn 2-[1-acetyl-3-[(E)-2-pyridin-2-ylethenyl]indazol-6-yl]sulfanyl-N-methylbenzamide
SMIL CNC(=O)c1ccccc1Sc2ccc3c(\C=C\c4ccccn4)nn(C(=O)C)c3c2
Tekniske data
Kemisk navn eller materiale N-Acetyl Axitinib
Analyt- eller komponentnavne N-Acetyl Axitinib
Forsendelsestilstand Room Temperature
InChI formel InChI=1S/C24H20N4O2S/c1-16(29)28-22-15-18(31-23-9-4-3-8-20(23)24(30)25-2)11-12-19(22)21(27-28)13-10-17-7-5-6-14-26-17/h3-15H,1-2H3,(H,25,30)/b13-10+
Løsningstype Neat
Industry Type Pharmaceutical
Mængde 2.5 mg