Få mere at vide
N-(6-Aminohexyl)-N'-(4-chlorophenyl)imidodicarbonimidic Diamide Dihydrochloride, TRC
Artikelnummer. 30133177
Skift visning
Mængde:
10 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133177 250 mg
30135783 50 mg
17993085 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30133177 Leverandør Toronto Research Chemicals Leverandørnr. TRCA610050250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 165678-61-5
Molekylær formel C14H25Cl3N6
Molekylvægt (g/mol) 383.75
Synonym Imidodicarbonimidic diamide, N-(6-aminohexyl)-N'-(4-chlorophenyl)-, hydrochloride (1:2), Imidodicarbonimidic diamide, N-(6-aminohexyl)-N'-(4-chlorophenyl)-, dihydrochloride (9CI), 1-(6-Aminohexyl)-5-(4-chlorophenyl)biguanide Dihydrochloride
IUPAC navn 1-[amino-(4-chloroanilino)methylidene]-2-(6-aminohexyl)guanidine
SMIL NCCCCCCNC(NC(NC1=CC=C(Cl)C=C1)=N)=N
Tekniske data
Kemisk navn eller materiale N-(6-Aminohexyl)-N'-(4-chlorophenyl)imidodicarbonimidic Diamide Dihydrochloride
Analyt- eller komponentnavne N-(6-Aminohexyl)-N'-(4-chlorophenyl)imidodicarbonimidic Diamide Dihydrochloride
Renhedsgrad noter HPLC
Forsendelsestilstand Room Temperature
InChI formel InChI=1S/C14H23ClN6/c15-11-5-7-12(8-6-11)20-14(18)21-13(17)19-10-4-2-1-3-9-16/h5-8H,1-4,9-10,16H2,(H5,17,18,19,20,21)
Løsningstype Neat
Industry Type Pharmaceutical
Mængde 250 mg
Formel vægt 382.1206
Procent renhed >95%