Få mere at vide
N-(4-Phenoxyphenyl)thiourea, 96%, Thermo Scientific™
Artikelnummer. 11352627
Skift visning
Mængde:
1 g
5 g
Pakningsstørrelse:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
11352627 5 g 5g
11342627 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 11352627 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. L11804.06
Request Bulk or Custom Format
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 76839-21-9
Molekylær formel C13H12N2OS
Molekylvægt (g/mol) 244.31
MDL nummer MFCD00041185
InChI nøgle SSZWMEWZRURHTK-UHFFFAOYSA-N
Synonym 1-4-phenoxyphenyl-2-thiourea, 4-phenoxyphenyl thiourea, n-4-phenoxyphenyl thiourea, 1-4-phenoxyphenyl thiourea, amino 4-phenoxyphenyl amino methane-1-thione, 1-4-phenoxyphenyl 2-thiourea, thiourea,n-4-phenoxyphenyl, thiourea, n-4-phenoxyphenyl
PubChem CID 2760349
IUPAC navn (4-phenoxyphenyl)thiourea
SMIL NC(=S)NC1=CC=C(OC2=CC=CC=C2)C=C1
Tekniske data
Kemisk navn eller materiale N-(4-Phenoxyphenyl)thiourea
Smeltepunkt 175°C to 177°C
Mængde 5 g
Beilstein 3309384
Formel vægt 244.32
Procent renhed 96%