Få mere at vide
N-[4-Cyano-3-(trifluoromethyl)phenyl]-2-methyl-2-oxiranecarboxamide, TRC
Artikelnummer. 30135163
Skift visning
Mængde:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135163 500 mg
30007150 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30135163 Leverandør Toronto Research Chemicals Leverandørnr. TRCC987585500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 90357-51-0
Molekylær formel C12 H9 F3 N2 O2
Molekylvægt (g/mol) 270.21
Synonym N-[4-Cyano-3-(trifluoromethyl)phenyl]-2-methyl-2-oxiranecarboxamide, Oxiranecarboxamide, N-[4-cyano-3-(trifluoromethyl)phenyl]-2-methyl- (9CI), Oxiranecarboxamide, N-[4-cyano-3-(trifluoromethyl)phenyl]-2-methyl-, (±)-, N-[4-Cyano-3-(trifluoromethyl)phenyl]-2-methyl-2-oxiranecarboxamide, 2-Methyloxirane-2-carboxylic acid N-(4-cyano-3-trifluoromethylphenyl)amide
IUPAC navn N-[4-cyano-3-(trifluoromethyl)phenyl]-2-methyloxirane-2-carboxamide
SMIL CC1(CO1)C(=O)Nc2ccc(C#N)c(c2)C(F)(F)F
Tekniske data
Kemisk navn eller materiale 1,2-Epoxy-2-methyl-N-[4-cyano-3-(trifluoromethyl)-phenyl]propionamide
Mængde 500 mg
InChI formel InChI=1S/C12H9F3N2O2/c1-11(6-19-11)10(18)17-8-3-2-7(5-16)9(4-8)12(13,14)15/h2-4H,6H2,1H3,(H,17,18)
Formel vægt 270.0616