Få mere at vide
Myrtenal (Mixture of Enantiomers), TRC
Artikelnummer. 30133728
Skift visning
Mængde:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133728 1 g
30134747 500 mg
30010111 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30133728 Leverandør Toronto Research Chemicals Leverandørnr. TRCM8850531G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 564-94-3
Molekylær formel C10H14O
Molekylvægt (g/mol) 150.22
Synonym 6,6-Dimethylbicyclo[3.1.1]hept-2-ene-2-carboxaldehyde, (±)-Myrtenal, 6,6-Dimethyl-2-norpinene-2-carboxaldehyde, Benihinal, Myrtenal, NSC 54384
IUPAC navn 6,6-dimethylbicyclo[3.1.1]hept-3-ene-4-carbaldehyde
SMIL CC1(C)C2CC=C(C=O)C1C2
Tekniske data
Kemisk navn eller materiale Myrtenal (Mixture of Enantiomers)
Mængde 1 g
InChI formel InChI=1S/C10H14O/c1-10(2)8-4-3-7(6-11)9(10)5-8/h3,6,8-9H,4-5H2,1-2H3
Formel vægt 150.1045