Få mere at vide
Mupirocin Lithium, TRC
Artikelnummer. 30474273
Skift visning
Mængde:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30474273 100 mg
30470808 1 g
30467089 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30474273 Leverandør Toronto Research Chemicals Leverandørnr. TRCM794010100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 73346-79-9
Molekylær formel C26 H43 O9 . Li
Molekylvægt (g/mol) 506.56
Synonym Mupirocin Lithium, BRL-4910B, Bactroban related
IUPAC navn lithium;9-[(E)-4-[(2S,3R,4R,5S)-3,4-dihydroxy-5-[[(2S,3S)-3-[(2S,3S)-3-hydroxybutan-2-yl]oxiran-2-yl]methyl]oxan-2-yl]-3-methylbut-2-enoyl]oxynonanoate
SMIL [Li+].C[C@H](O)[C@H](C)[C@@H]1O[C@H]1C[C@H]2CO[C@@H](C\C(=C\C(=O)OCCCCCCCCC(=O)[O-])\C)[C@H](O)[C@@H]2O
Tekniske data
Kemisk navn eller materiale Mupirocin Lithium
Mængde 100 mg
InChI formel InChI=1S/C26H44O9.Li/c1-16(13-23(30)33-11-9-7-5-4-6-8-10-22(28)29)12-20-25(32)24(31)19(15-34-20)14-21-26(35-21)17(2)18(3)27;/h13,17-21,24-27,31-32H,4-12,14-15H2,1-3H3,(H,28,29);/q;+1/p-1/b16-13+;/t17-,18-,19-,20-,21-,24+,25-,26-;/m0./s1
Formel vægt 506.3067