missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Biosciences™ Multi-Check Control (CE/IVD)
Intended as a complete process control for immunophenotyping by flow cytometry.
2035.00 DKK - 5175.00 DKK
Specifications
Certifications/Compliance | IVD 2/2014, CE 23-5203-04 |
---|---|
Content And Storage | Store vials upright, tightly capped, at 2°C to 8°C. |
Stability | Unopened vials are stable until the expiration date. Opened vials are stable for 9 thermal cycles when handled properly. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15845938
|
BD Biosciences™
340911 |
1 x 2.5 mL |
N/A
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15855938
|
BD Biosciences™
340912 |
2 x 2.5 mL |
2035.00 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15865938
|
BD Biosciences™
340913 |
5 x 2.5 mL |
5175.00 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Complete process control for immunophenotyping by flow cytometry.
- Control for antibody staining, red blood cell (RBC) lysis, instrument setup and performance, and data analysis.
- Stable control with assigned values that can be used to monitor the immunophenotyping process.
- The BD Multi-Check Control should be treated in the same manner as whole blood.
Specifications
IVD 2/2014, CE 23-5203-04 | |
Store vials upright, tightly capped, at 2°C to 8°C. | |
Unopened vials are stable until the expiration date. Opened vials are stable for 9 thermal cycles when handled properly. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BD Biosciences™ Multi-Check Control (CE/IVD)