Learn More
BD Biosciences™ Multi-Check Control (CE/IVD)
Intended as a complete process control for immunophenotyping by flow cytometry.
Brand: BD Biosciences™ 340912
This item is not returnable.
View return policy
Description
- Complete process control for immunophenotyping by flow cytometry.
- Control for antibody staining, red blood cell (RBC) lysis, instrument setup and performance, and data analysis.
- Stable control with assigned values that can be used to monitor the immunophenotyping process.
- The BD Multi-Check Control should be treated in the same manner as whole blood.
Specifications
IVD 2/2014, CE 23-5203-04 | |
Store vials upright, tightly capped, at 2°C to 8°C. | |
2 x 2.5 mL | |
Unopened vials are stable until the expiration date. Opened vials are stable for 9 thermal cycles when handled properly. |
Product Content Correction
Product Title
BD Biosciences™ Multi-Check Control (CE/IVD) > 2 x 2.5 mL
Spot an opportunity for improvement?Share a Content Correction