Få mere at vide
MUC13 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55487-1-AP-20UL
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
The MUC13 antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human MUC13. This antibody recognizes human antigen. The MUC13 antibody has been validated for the following applications: WB, IHC, ELISA analysis.Tekniske data
| MUC13 | |
| Polyclonal | |
| Unconjugated | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human | |
| 90 kDa | |
| Liquid |
| Western Blot, Immunohistochemistry, ELISA | |
| 1500 μg/mL | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 56667 | |
| Store at -20°C. Stable for one year after shipment. | |
| Polyclonal Antibody | |
| IgG |