Få mere at vide
MS023 dihydrochloride, MedChemExpress
Artikelnummer. 16497646
Encompass_Preferred
Skift visning
Mængde:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Pakningsstørrelse:
10mg
25mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16497646 100 mg -
16487818 10 mg 10mg
16433777 50 mg -
16430288 5 mg -
16423666 25 mg 25mg
16420326 2 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Artikelnummer. 16497646 Leverandør Medchem Express Leverandørnr. HY19615B
MedChemExpress MS023 dihydrochloride is a potent, selective, and cell-active inhibitor of human type I protein arginine methyltransferases (PRMTs) inhibitor, with IC50s of 30, 119, 83, 4 and 5 nM for PRMT1, PRMT3, PRMT4, PRMT6, and PRMT8, respectively.
MS023 dihydrochloride is a potent, selective, and cell-active inhibitor of human type I protein arginine methyltransferases (PRMTs) inhibitor, with IC50s of 30, 119, 83, 4 and 5 nM for PRMT1, PRMT3, PRMT4, PRMT6, and PRMT8, respectively.
Tekniske data
Kemisk navn eller materiale MS023 dihydrochloride
CAS 1992047-64-9
Farve Pale Purple
Renhedsgrad noter Research
Molekylær formel C17H27Cl2N3O
Mængde 100 mg
Opløselighedsinformation DMSO : ≥ 150 mg/mL (416.30 mM) ∣H2O : 33.33 mg/mL (92.50 mM; Need ultrasonic)
SMIL CC(OC1=CC=C(C=C1)C2=CNC=C2CN(C)CCN)C.Cl.Cl
Molekylvægt (g/mol) 360.32
Formel vægt 360.32
Procent renhed 99.78%
Grad Research
Fysisk form Powder
Til brug med (applikation) Cancer-programmed cell death