Få mere at vide
Tocris Bioscience™ MRK 560
GREENER_CHOICE
Artikelnummer. 18015452
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18015452 10 mg
18005112 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18015452 Leverandør Tocris Bioscience™ Leverandørnr. 4000/10
γ-secretase inhibitor; attenuates amyloid plaque deposition
γ-secretase inhibitor; inhibits proteolytic cleavage of amyloid precursor protein (APP) over the Notch pathway. Reduces levels of Aβ in the brain (inhibits Aβ40 and Aβ42 in SH-SY5Y neuroblastoma cells with an IC50 of 0.65 nM); attenuates plaque deposition. Orally bioavailable.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 677772-84-8
Molekylær formel C19H17ClF5NO4S2
Tekniske data
Kemisk navn eller materiale MRK 560
Mængde 10 mg
Formel vægt 517.92
Renhed >99%
Molekylvægt (g/mol) 517.91