Learn More
MP Biomedicals™ Nisin
Nisin content: 2.5% Remainder is NaCl and milk solids (denatured)
1065.30 DKK - 3916.70 DKK
Specifications
Content And Storage 15° to 30°C
pH 3.31
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11495041
View Documents Promotion Details
MP Biomedicals™
0215583905
5 g
1065.30 DKK
5g
View alternative products
Save Up to 
»
Estimated Shipment: 18-07-2024
to see stock.
11405051
View Documents Promotion Details
MP Biomedicals™
0215583925
25 g
3916.70 DKK
25g
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Description
Description
  • Molecular Formula: C14H230N42O37S7
  • Molecular Weight: 3354.3
Specifications
Specifications
15° to 30°C
3.31