missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Nisin

Nisin content: 2.5% Remainder is NaCl and milk solids (denatured)

Brand:  MP Biomedicals™ 0215583925

Additional Details : CAS Number : 1414-45-5 Weight : 0.02500kg

 View more versions of this product

Product Code. 11405051

  • 3916.70 DKK / 25g
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Molecular Formula: C14H230N42O37S7
  • Molecular Weight: 3354.3

Specifications

Specifications

15° to 30°C
3.31
25 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers