Learn More
MP Biomedicals™ Casein Hydrolysate Acid
Sulfuric acid digest of casein
1611.20 DKK - 3275.40 DKK
Specifications
Content And Storage 15° to 30°C
Product Type Amicase
View More Specs
Products 2
Product Code Brand pH Quantity Price Quantity & Availability  
Product Code Brand pH Quantity Price Quantity & Availability  
11426131
View Documents Promotion Details
MP Biomedicals™
0219102301
5 to 7 1 lb.
1611.20 DKK
1lb
View alternative products
Save Up to 
»
Estimated Shipment: 22-05-2024
to see stock.
11493230
View Documents Promotion Details
MP Biomedicals™
0210129105
5 to 7 5 lb.
3275.40 DKK
5lb
View alternative products
Save Up to 
»
Estimated Shipment: 22-05-2024
to see stock.
Description
Description
Lower salt content than the hydrochloric acid digest. Essentially tryptophan free; 5.0 pH (2% soln.). Nitrogen approx. 13.0%; amino nitrogen approx. 9.2%.
Specifications
Specifications
15° to 30°C
Amicase