Learn More
MP Biomedicals™ Casein Hydrolysate Acid
Sulfuric acid digest of casein
Brand: MP Biomedicals™ 0210129105
Additional Details : Weight : 2.27000kg
Description
Lower salt content than the hydrochloric acid digest. Essentially tryptophan free; 5.0 pH (2% soln.). Nitrogen approx. 13.0%; amino nitrogen approx. 9.2%.
Specifications
15° to 30°C | |
Amicase | |
5 to 7 | |
5 lb. |