missing translation for 'onlineSavingsMsg'
Få mere at vide

Montelukast Sulfone, TRC

Artikelnummer. 30468846
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30468846 10 mg
30466546 5 mg
30475909 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30468846 Leverandør Toronto Research Chemicals Leverandørnr. TRCM56803510MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 1266620-74-9
Molekylær formel C35 H36 Cl N O5 S
Molekylvægt (g/mol) 618.18
Synonym 1-[[[(1R)-1-[3-[(1E)-2-(7-Chloro-2-quinolinyl)ethenyl]phenyl]-3-[2-(1-hydroxy-1-methylethyl)phenyl]propyl]sulfonyl]methyl]cyclopropaneacetic Acid, Montelukast Sulfone
IUPAC navn 2-[1-[[(1R)-1-[3-[(E)-2-(7-chloroquinolin-2-yl)ethenyl]phenyl]-3-[2-(2-hydroxypropan-2-yl)phenyl]propyl]sulfonylmethyl]cyclopropyl]acetic acid
SMIL CC(C)(O)c1ccccc1CC[C@H](c2cccc(\C=C\c3ccc4ccc(Cl)cc4n3)c2)S(=O)(=O)CC5(CC(=O)O)CC5

Tekniske data

Kemisk navn eller materiale Montelukast Sulfone
Mængde 10 mg
InChI formel InChI=1S/C35H36ClNO5S/c1-34(2,40)30-9-4-3-7-25(30)13-17-32(43(41,42)23-35(18-19-35)22-33(38)39)27-8-5-6-24(20-27)10-15-29-16-12-26-11-14-28(36)21-31(26)37-29/h3-12,14-16,20-21,32,40H,13,17-19,22-23H2,1-2H3,(H,38,39)/b15-10+/t32-/m1/s1
Formel vægt 617.2003
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.