Få mere at vide
Mono-sec-butyl Phthalate, TRC
Artikelnummer. 30466243
Skift visning
Mængde:
250 mg
2.5 g
Pakningsstørrelse:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30466243 250 mg -
30472132 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30466243 Leverandør Toronto Research Chemicals Leverandørnr. TRCM525110250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 53623-59-9
Molekylær formel C12H14O4
Molekylvægt (g/mol) 222.24
Synonym 1,2-Benzenedicarboxylic Acid 1-(1-methylpropyl) Ester, 1,2-Benzenedicarboxylic Acid Mono(1-methylpropyl) Ester, Phthalic Acid sec-Butyl Ester, NSC 29059
IUPAC navn 2-butan-2-yloxycarbonylbenzoic acid
SMIL CCC(C)OC(=O)c1ccccc1C(=O)O
Tekniske data
Kemisk navn eller materiale Mono-sec-butyl Phthalate
Mængde 250 mg
InChI formel InChI=1S/C12H14O4/c1-3-8(2)16-12(15)10-7-5-4-6-9(10)11(13)14/h4-8H,3H2,1-2H3,(H,13,14)
Formel vægt 222.0892