Få mere at vide
Mono-7-carboxyheptyl Phthalate, TRC
Artikelnummer. 30009138
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30009138 25 mg
30005655 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30009138 Leverandør Toronto Research Chemicals Leverandørnr. TRCM52558025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 856869-57-3
Molekylær formel C16H20O6
Molekylvægt (g/mol) 308.33
Synonym Mono-7-carboxyheptyl phthalate, 2-{[(7-carboxyheptyl)oxy]carbonyl}benzoic acid, 1,2-benzenedicarboxylic acid, mono-7-carboxyheptyl ester
IUPAC navn 2-(7-carboxyheptoxycarbonyl)benzoic acid
SMIL C1=CC=C(C(=C1)C(=O)O)C(=O)OCCCCCCCC(=O)O
Tekniske data
Kemisk navn eller materiale Mono(7-carboxyheptyl) phthalate
Mængde 25 mg
InChI formel InChI=1S/C16H20O6/c17-14(18)10-4-2-1-3-7-11-22-16(21)13-9-6-5-8-12(13)15(19)20/h5-6,8-9H,1-4,7,10-11H2,(H,17,18)(H,19,20)
Formel vægt 308.126