Få mere at vide
Tocris Bioscience™ MONNA, >98%
GREENER_CHOICE
Artikelnummer. 30000967
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30000967 10 mg
30000944 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30000967 Leverandør Tocris Bioscience™ Leverandørnr. 5770/10
Potent TMEM16A (Anoctamin-1) blocker
MONNA is a potent TMEM16A (Anoctamin-1) blocker (IC50 = 80 nM). Exhibits >100-fold selectivity for TMEM16A over bestrophin-1, CLC2 and CFTR. Induces vasorelaxation of rodent resistance arteries in presence or absence of chloride ions. Carbachol (Cat. No. 2810) induced contraction of bronchial myocytes is reduced by MONNA and intracellular calcium release is increased.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1572936-83-4
Molekylær formel C18H14N2O5
Tekniske data
Kemisk navn eller materiale 2-[(4-Methoxy-2-naphthalenyl)amino]-5-nitro-benzoic acid
Mængde 10 mg
Opløselighed Soluble to 100mM in DMSO
Formel vægt 338.31
Renhed >98%
Molekylvægt (g/mol) 338.319