Få mere at vide
Mometasone Furoate Impurity A, TRC
Artikelnummer. 30135344
Skift visning
Mængde:
2.5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135344 25 mg
30138346 10 mg
17980745 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30135344 Leverandør Toronto Research Chemicals Leverandørnr. TRCM49010025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 83880-65-3
Molekylær formel C27 H29 Cl O5
Molekylvægt (g/mol) 468.97
Synonym Pregna-1,4,9(11)-triene-3,20-dione, 21-chloro-17-[(2-furanylcarbonyl)oxy]-16-methyl-, (16α)-, 21-Chloro-16α-methyl-3,20-dioxopregna-1,4,9(11)-trien-17-yl Furan-2-carboxylate, Mometasone Furoate Imp. A (EP)
IUPAC navn [(8S,10S,13S,14S,16R,17R)-17-(2-chloroacetyl)-10,13,16-trimethyl-3-oxo-7,8,12,14,15,16-hexahydro-6H-cyclopenta[a]phenanthren-17-yl] furan-2-carboxylate
SMIL C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)C3=CC[C@]2(C)[C@@]1(OC(=O)c5occc5)C(=O)CCl
Tekniske data
Kemisk navn eller materiale 21-Chloro-16alpha-methyl-3,20-dioxopregna-1,4,9(11)-trien-17-yl Furan-2-carboxylate
Mængde 25 mg
InChI formel InChI=1S/C27H29ClO5/c1-16-13-21-19-7-6-17-14-18(29)8-10-25(17,2)20(19)9-11-26(21,3)27(16,23(30)15-28)33-24(31)22-5-4-12-32-22/h4-5,8-10,12,14,16,19,21H,6-7,11,13,15H2,1-3H3/t16-,19-,21+,25+,26+,27+/m1/s1
Formel vægt 468.1704