Få mere at vide
Thermo Scientific™ Molecular BioProducts Snap-Cap Plain Centrifuge Tubes
Brand: Thermo Scientific™ 3439G
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
- With flat cap
- No frosting or graduations
- Certified RNase/DNase, RNA/DNA, ATP and pyrogen free
Tekniske data
Flat | |
0.6 mL | |
Polypropylene | |
Bag | |
No |
RNase, DNase, RNA, DNA, ATP, pyrogen-free | |
Green | |
Flat cap | |
Sterile | |
Flat Cap |
Korrektion af produktindhold
Produkttitel
Thermo Scientific™ Molecular BioProducts Snap-Cap Plain Centrifuge Tubes > Sterile; Green; 0.6mL
Ser du en mulighed for forbedring?Del en indholdskorrektion