missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
MMP-24/MT5-MMP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP1-59458
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
MMP-24/MT5-MMP Polyclonal specifically detects MMP-24/MT5-MMP in Human samples. It is validated for Western Blot.
Especificaciones
| MMP-24/MT5-MMP | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| EC 3.4.24, EC 3.4.24.-, EC 3.4.24.80, matrix metallopeptidase 24 (membrane-inserted), matrix metalloproteinase 24 (membrane-inserted), matrix metalloproteinase-24, membrane-type 5 matrix metalloproteinase, Membrane-type matrix metalloproteinase 5, Membrane-type-5 matrix metalloproteinase, MMP-24, MT5MMP, MT5-MMPMMP25, MT-MMP 5, MTMMP5, MT-MMP5 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 10893 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q9Y5R2 | |
| MMP24 | |
| Synthetic peptides corresponding to MMP24(matrix metallopeptidase 24 (membrane-inserted)) The peptide sequence was selected from the middle region of MMP24 (NP_006681). Peptide sequence GSCLPREGIDTALRWEPVGKTYFFKGERYWRYSEERRATDPGYPKPITVW. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Bovine: 100%; Canine: 100%; Guinea pig: 100%; Equine: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Chicken: 85%; Zebrafish: 85%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido