missing translation for 'onlineSavingsMsg'
Få mere at vide

ML141, MedChemExpress

Artikelnummer. 16485318
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
10 mg
50 mg
100 mg
Pakningsstørrelse:
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16485318 10 mg -
16475966 100 mg -
16438037 5 mg -
16431527 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16485318 Leverandør Medchem Express Leverandørnr. HY12755

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress ML141 (CID-2950007) is a potent, allosteric, selective and reversible non-competitive inhibitor of Cdc42 GTPase. ML141 inhibits Cdc42 wild type and Cdc42 Q61L mutant with EC50s of 2.1 and 2.6 μM, respectively. ML141 shows low micromolar potency and selectivity against other members of the Rho family of GTPases (Rac1, Rab2, Rab7). ML141 do not show cytotoxicity in multiple cell lines.

ML141 (CID-2950007) is a potent, allosteric, selective and reversible non-competitive inhibitor of Cdc42 GTPase. ML141 inhibits Cdc42 wild type and Cdc42 Q61L mutant with EC50s of 2.1 and 2.6 μM, respectively. ML141 shows low micromolar potency and selectivity against other members of the Rho family of GTPases (Rac1, Rab2, Rab7). ML141 do not show cytotoxicity in multiple cell lines.

Tekniske data

Kemisk navn eller materiale ML141
CAS 71203-35-5
Farve Off-White
Renhedsgrad noter Research
Molekylær formel C22H21N3O3S
Mængde 10 mg
Synonym CID-2950007
Opløselighedsinformation DMSO : ≥ 55 mg/mL (134.97 mM)
SMIL O=S(C1=CC=C(N2N=C(C3=CC=CC=C3)CC2C4=CC=C(OC)C=C4)C=C1)(N)=O
Molekylvægt (g/mol) 407.49
Formel vægt 407.49
Procent renhed 98.03%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-Kinase/protease
Vis mere Vis mindre

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.