missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MKNK2 (MNK2) Human Protein
MKNK1 and MKNK2 are the Map kinase-interacting kinases.
Brand: Invitrogen™ PV5608
Additional Details : Weight : 0.01000kg
Description
The Map kinase-interacting kinases are activated by Map kinases, regulate protein synthesis by phosphroylation eIF4E on Ser209, and may contribute to tumorigenesis.Specifications
NP_060042.2 | |
50mM Tris pH 7.5, 150mM NaCl, 0.02% Trition X-100, 0.5mM EDTA, 2mM DTT, 50% Glycerol. | |
1mg | |
RUO - research use only | |
MKNK2 (MNK2) | |
Human | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
74.2kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
MAPKAPK Kinase Family | |
GST Tag | |
Frozen |