missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MKNK2 (MNK2) Human Protein
MKNK1 and MKNK2 are the Map kinase-interacting kinases.
4380.00 DKK - 57500.00 DKK
Specifications
Accession Number | NP_060042.2 |
---|---|
Formulation | 50mM Tris pH 7.5, 150mM NaCl, 0.02% Trition X-100, 0.5mM EDTA, 2mM DTT, 50% Glycerol. |
Molecular Weight (g/mol) | 74.2kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Structural Form | Full Length |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10582974
|
Invitrogen™
PV5607 |
10μg |
4380.00 DKK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15478643
|
Invitrogen™
PR8046A |
100μg |
13290.00 DKK
100µg |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15428793
|
Invitrogen™
PV5608 |
1mg |
57500.00 DKK
1mg |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Map kinase-interacting kinases are activated by Map kinases, regulate protein synthesis by phosphroylation eIF4E on Ser209, and may contribute to tumorigenesis.Specifications
NP_060042.2 | |
74.2kDa | |
Full Length | |
MAPKAPK Kinase Family | |
GST Tag | |
Frozen |
50mM Tris pH 7.5, 150mM NaCl, 0.02% Trition X-100, 0.5mM EDTA, 2mM DTT, 50% Glycerol. | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
MKNK2 (MNK2) | |
Human | |
Baculovirus |