Få mere at vide
MK-886, TRC
Artikelnummer. 30469717
Skift visning
Mængde:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30469717 10 mg
30478488 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30469717 Leverandør Toronto Research Chemicals Leverandørnr. TRCM42505310MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 118414-82-7
Molekylær formel C27H34ClNO2S
Molekylvægt (g/mol) 472.08
Synonym MK-886, L 663536
SMIL CC(C)c1ccc2c(c1)c(SC(C)(C)C)c(CC(C)(C)C(=O)O)n2Cc3ccc(Cl)cc3
Tekniske data
Kemisk navn eller materiale 1-[(4-Chlorophenyl)methyl]-3-[(1,1-dimethylethyl)thio]-alpha,alpha-dimethyl-5-(1-methylethyl)-1H-indole-2-propanoic Acid