Learn More
Merck Microsyringe Filter Holder, 25 mm
Accessories for filter holders for sample preparation.
3435.00 DKK - 4335.00 DKK
Specifications
Autoclavable | Autoclavable |
---|---|
Disposable | Reusable |
For Use With (Equipment) | Syringe Filter Holder |
Material | Stainless Steel |
Sterility | Non-sterile |
Product Code | Brand | Inlet Style | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Inlet Style | Price | Quantity & Availability | ||||||
10469142
|
Merck
XX3002514 |
1/4 in. NPTF, Stainless Steel |
4335.00 DKK
Each |
Estimated Shipment: 28-07-2025
Log in to see stock. |
||||||
10492321
|
Merck
XX3002500 |
Female Luer-Lok™, Stainless Steel |
3435.00 DKK
Each |
Estimated Shipment: 28-07-2025
Log in to see stock. |
||||||
Description
Accessories for filter holders for sample preparation.
Make your own syringe filter. Microsyringe filter holders give you the flexibility to select the membrane type and diameter that are most appropriate for your application. For sterile filtration, autoclave your filter holders with the membrane in place. Ultraclean or sterilize small volumes of liquids dispensed by a syringe or install in-line to filter larger volumes.
Features and Benefits:
- Durable stainless steel construction ensures years of use, decreasing waste from disposable syringe filters.
- Autoclavable with filter in place for sterile filtration.
Specifications
Autoclavable | |
Syringe Filter Holder | |
Non-sterile | |
Male Luer-Slip | |
Filter Holder | |
3.2 cm |
Reusable | |
Stainless Steel | |
3.9 cm2 | |
6.9 bar (100 psi) | |
1.3 in. |