Få mere at vide
IBA Lifesciences MHC I-Strep H-2 Kb; Ovalbumin (SIINFEKL)
GREENER_CHOICE
Artikelnummer. 15623908 Shop alle IBA Lifesciences produkter
Mængde:
40 μL
Pakningsstørrelse:
40µL
Artikelnummer. 15623908
Brand: IBA Lifesciences 67015001
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
IBA Lifesciences GmbH MHC I-Strep H-2 Kb Ovalbumin SIINFEKL is for fluorescent staining/FACS as well as for magnetic isolation of antigen-specific (CD8+) T cells.
  • MHC I-Strep H-2 Kb; Ovalbumin (SIINFEKL) is a recombinantly expressed MHC class I molecule that presents a peptide from a Ovalbumin.
  • Our MHC I Streptamer™ approach is based on our Strep-tag technology and facilitates reversible antigen-specific T cell labeling.
  • A Twin-Strep-tag™ fused to the MHC I-Strep permits multimerization of MHC I complexes on a Strep-Tactin™ backbone, resulting in the formation of MHC I Streptamers.
  • For multimerizing MHC I-Streps, Strep-Tactin™ TACS Agarose columns, Strep-Tactin™ Magnetic Microbeads or fluorophore-conjugated Strep-Tactin™ can be used.
  • This allows affinity chromatographic or magnetic isolation or cell staining for flow cytometry analysis and sorting (FACS), depending on the experimental requirements.
  • The addition of biotin reverses the multimerization of MHC class I molecules, leading to their dissociation from the target cells.
  • The resulting label-free cells are highly viable and fully functional.
  • Tekniske data
    Kapacitet (metrisk) 40 μL
    Sættets indhold Fluorescent reagent for staining, magnetic T cell labeling and isolation
    Mængde 40 μL