Få mere at vide
Metribuzin, TRC
Artikelnummer. 30005696
Skift visning
Mængde:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30005696 500 mg
17943915 1 g
17994895 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30005696 Leverandør Toronto Research Chemicals Leverandørnr. TRCM338920500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 21087-64-9
Molekylær formel C8 H14 N4 O S
Molekylvægt (g/mol) 214.29
Synonym 1,2,4-Triazin-5(4H)-one, 4-amino-6-(1,1-dimethylethyl)-3-(methylthio)-, as-Triazin-5(4H)-one, 4-amino-6-tert-butyl-3-(methylthio)- (8CI), 4-Amino-6-(1,1-dimethylethyl)-3-(methylthio)-1,2,4-triazin-5(4H)-one, 3-Methylthio-4-amino-6-tert-butyl-1,2,4-triazin-5(4H)-one, 3-Methylthio-4-amino-6-tert-butyl-1,2,4-triazin-5-one, 4-Amino-6-tert-butyl-3-(methylthio)-1,2,4-triazin-5(4H)-one, 4-Amino-6-tert-butyl-3-(methylthio)-1,2,4-triazin-5-one, 4-Amino-6-tert-butyl-3-(methylthio)-1,2,4-triazine-5(4H)-one, 4-Amino-6-tert-butyl-3-(methylthio)-4,5-dihydro-1,2,4-triazin-5-one, 4-Amino-6-tert-butyl-3-(methylthio)-as-triazin-5(4H)-one, 4-Amino-6-tert-butyl-4,5-dihydro-3-methylthio-1,2,4-triazin-5-one, BAY 6159, BAY 61597, BAY 6159H, BAY 94337, Bayer 6159, DIC 1468, DPX 2504, Lexone, Lexone DF, Metribuzin, Metribuzine, Sencor, Sencor 480, Sencor 4F, Sencor 75DF, Sencor Extra, Sencorex, Sencorex L.F., Senkor, Soccer, Soccer (herbicide), Tribute, Tribute (metribuzin herbicide), Zontran
IUPAC navn 4-amino-6-tert-butyl-3-methylsulfanyl-1,2,4-triazin-5-one
SMIL CSC1=NN=C(C(=O)N1N)C(C)(C)C
Tekniske data
Kemisk navn eller materiale Metribuzin
Mængde 500 mg
InChI formel InChI=1S/C8H14N4OS/c1-8(2,3)5-6(13)12(9)7(14-4)11-10-5/h9H2,1-4H3
Formel vægt 214.0888