Learn More
Methyle2-isothiocyanatobenzoate, 97%, Thermo Scientific™
186.00 DKK - 567.00 DKK
Chemical Identifiers
CAS | 16024-82-1 |
---|---|
Molecular Formula | C9H7NO2S |
Molecular Weight (g/mol) | 193.22 |
MDL Number | MFCD00041062 |
InChI Key | UNXVHBOJSCWVCD-UHFFFAOYSA-N |
Synonym | 2-isothiocyanato-benzoic acid methyl ester, 2-methoxycarbonylphenyl isothiocyanate, 2-methoxycarbonyl phenyl isothiocyanate, benzoic acid,2-isothiocyanato-, methyl ester, benzoic acid, 2-isothiocyanato-, methyl ester, acmc-209dkr, methyl2-isothiocyanatobenzoate, methyl 2-isothiocyanato benzoate, methyl 2-isothiocyanatobenzenecarboxylate, 2-isothiocyanatobenzoic acid methyl esterShow More |
PubChem CID | 85243 |
IUPAC Name | methyl 2-isothiocyanatobenzoate |
SMILES | COC(=O)C1=CC=CC=C1N=C=S |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10302315
|
Thermo Scientific Maybridge
SB01695DE |
5 g |
186.00 DKK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10743574
|
Thermo Scientific Maybridge
SB01695EA |
10 g |
296.00 DKK
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10722037
|
Thermo Scientific Maybridge
SB01695EB |
25 g |
567.00 DKK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
16024-82-1 | |
193.22 | |
UNXVHBOJSCWVCD-UHFFFAOYSA-N | |
85243 | |
COC(=O)C1=CC=CC=C1N=C=S |
C9H7NO2S | |
MFCD00041062 | |
2-isothiocyanato-benzoic acid methyl ester, 2-methoxycarbonylphenyl isothiocyanate, 2-methoxycarbonyl phenyl isothiocyanate, benzoic acid,2-isothiocyanato-, methyl ester, benzoic acid, 2-isothiocyanato-, methyl ester, acmc-209dkr, methyl2-isothiocyanatobenzoate, methyl 2-isothiocyanato benzoate, methyl 2-isothiocyanatobenzenecarboxylate, 2-isothiocyanatobenzoic acid methyl esterShow More | |
methyl 2-isothiocyanatobenzoate |
Specifications
16024-82-1 | |
1.2500g/mL | |
C9H7NO2S | |
MFCD00041062 | |
UNXVHBOJSCWVCD-UHFFFAOYSA-N | |
methyl 2-isothiocyanatobenzoate | |
85243 | |
97% |
97.0 | |
97% | |
SCNC6H4CO2CH3 | |
2-isothiocyanato-benzoic acid methyl ester, 2-methoxycarbonylphenyl isothiocyanate, 2-methoxycarbonyl phenyl isothiocyanate, benzoic acid,2-isothiocyanato-, methyl ester, benzoic acid, 2-isothiocyanato-, methyl ester, acmc-209dkr, methyl2-isothiocyanatobenzoate, methyl 2-isothiocyanato benzoate, methyl 2-isothiocyanatobenzenecarboxylate, 2-isothiocyanatobenzoic acid methyl ester | |
COC(=O)C1=CC=CC=C1N=C=S | |
193.22 | |
193.23 | |
Methyl 2-isothiocyanatobenzoate |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.