Få mere at vide
Methyl Stearidonate, TRC
Artikelnummer. 30480956
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30480956 10 mg
30466035 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30480956 Leverandør Toronto Research Chemicals Leverandørnr. TRCM32815010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 73097-00-4
Molekylær formel C19 H30 O2
Molekylvægt (g/mol) 290.4403
Synonym Methyl (all-cis)-6,9,12,15-octadecatetraenoate,(6Z,9Z,12Z,15Z)-Octadecatetraenoic acid methyl ester,(6Z,9Z,12Z,15Z)-Methyl octadeca-6,9,12,15-tetraenoate,(Z,Z,Z,Z)-6,9,12,15-OCTADECATETRAENOIC ACID METHYL ESTER
IUPAC navn methyl (6Z,9Z,12Z,15Z)-octadeca-6,9,12,15-tetraenoate
SMIL CC\C=C/C\C=C/C\C=C/C\C=C/CCCCC(=O)OC
Tekniske data
Kemisk navn eller materiale Methyl stearidonate
Mængde 10 mg
InChI formel InChI=1S/C19H30O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19(20)21-2/h4-5,7-8,10-11,13-14H,3,6,9,12,15-18H2,1-2H3/b5-4-,8-7-,11-10-,14-13-
Formel vægt 290.2246