missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl red, For ACS analysis, MP Biomedicals™
Shop All MP Biomedical Products
Click to view available options
Quantity:
250 g
Unit Size:
250g
Specifications
Specifications
Formula Weight | 269.3 |
Color | Red |
Quantity | 250 g |
Physical Form | Powder |
Chemical Name or Material | Methyl red, For ACS analysis |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 9
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Methyl red, For ACS analysis, MP Biomedicals™ >
Spot an opportunity for improvement?Share a Content Correction