Få mere at vide
Methyl 5-Chloropentanoate, TRC
Artikelnummer. 30476117
Skift visning
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30476117 100 mg
30472750 500 mg
30479696 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30476117 Leverandør Toronto Research Chemicals Leverandørnr. TRCM340460100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 14273-86-0
Molekylær formel C6H11O2Cl
Molekylvægt (g/mol) 150.6
Synonym Apixaban Impurity 84
IUPAC navn methyl 5-chloropentanoate
SMIL COC(=O)CCCCCl
Tekniske data
Kemisk navn eller materiale Methyl 5-Chloropentanoate
Mængde 100 mg
InChI formel InChI=1S/C6H11ClO2/c1-9-6(8)4-2-3-5-7/h2-5H2,1H3
Formel vægt 150.0448