Få mere at vide
Methyl 5-Bromo-4-Chloronicotinate, TRC
Artikelnummer. 30467577
Skift visning
Mængde:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30467577 1 g
30480621 500 mg
30465678 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30467577 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2945331G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1256808-62-4
Molekylær formel C7H5NO2ClBr
Molekylvægt (g/mol) 250.47
Synonym Methyl 5-Bromo-4-chloro-3-pyridinecarboxylate
IUPAC navn methyl 5-bromo-4-chloropyridine-3-carboxylate
SMIL COC(=O)c1cncc(Br)c1Cl
Tekniske data
Kemisk navn eller materiale Methyl 5-Bromo-4-Chloronicotinate
Mængde 1 g
InChI formel InChI=1S/C7H5BrClNO2/c1-12-7(11)4-2-10-3-5(8)6(4)9/h2-3H,1H3
Formel vægt 248.9192