Få mere at vide
Methyl 4-Hydroxy-2-methyl-2H-Thieno[2,3-e]-1,2-thiazine-3-carboxylic Acid 1,1-Dioxide Ester, TRC
Artikelnummer. 30011447
Skift visning
Mængde:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30011447 250 mg
30008655 50 mg
30006687 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30011447 Leverandør Toronto Research Chemicals Leverandørnr. TRCM312705250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 59804-25-0
Molekylær formel C9 H9 N O5 S2
Molekylvægt (g/mol) 275.3
Synonym Methyl 4-Hydroxy-2-methyl-2H-thieno[2,3-e]1,2-thiazine-3-carboxylate 1,1-Dioxide, Tenoxicam Imp. B (EP)
IUPAC navn methyl 4-hydroxy-2-methyl-1,1-dioxothieno[2,3-e]thiazine-3-carboxylate
SMIL COC(=O)C1=C(O)c2sccc2S(=O)(=O)N1C
Tekniske data
Kemisk navn eller materiale Methyl 4-Hydroxy-2-methyl-2H-thieno[2,3-e]1,2-thiazine-3-carboxylate 1,1-Dioxide
Mængde 250 mg
InChI formel InChI=1S/C9H9NO5S2/c1-10-6(9(12)15-2)7(11)8-5(3-4-16-8)17(10,13)14/h3-4,11H,1-2H3
Formel vægt 274.9922