CAS RN 59804-25-0
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
Methyl 4-Hydroxy-2-methyl-2H-thieno[2,3-e]1,2-thiazine-3-carboxylate 1,1-Dioxide, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Methyl 4-Hydroxy-2-methyl-2H-Thieno[2,3-e]-1,2-thiazine-3-carboxylic Acid 1,1-Dioxide Ester, TRC
CAS: 59804-25-0 Molekylær formel: C9 H9 N O5 S2 Molekylvægt (g/mol): 275.3 Synonym: Methyl 4-Hydroxy-2-methyl-2H-thieno[2,3-e]1,2-thiazine-3-carboxylate 1,1-Dioxide,Tenoxicam Imp. B (EP) IUPAC navn: methyl 4-hydroxy-2-methyl-1,1-dioxothieno[2,3-e]thiazine-3-carboxylate SMIL: COC(=O)C1=C(O)c2sccc2S(=O)(=O)N1C