Få mere at vide
Methyl (3R)-3-(tert-Butyldimethylsilanyloxy)-5-oxo-6-(triphenylphosphanylidene)hexanoate, TRC
Artikelnummer. 30467276
Skift visning
Mængde:
10 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30467276 100 g
30483418 25 g
30478787 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30467276 Leverandør Toronto Research Chemicals Leverandørnr. TRCM288100100G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 147118-35-2
Molekylær formel C31 H39 O4 P Si
Molekylvægt (g/mol) 534.7
Synonym Hexanoic acid, 3-[[(1,1-dimethylethyl)dimethylsilyl]oxy]-5-oxo-6-(triphenylphosphoranylidene)-, methyl ester, (R)-, (3R)-3-(tert-Butyldimethylsilanyloxy)-5-oxo-6-(triphenylphosphanylidene)hexanoic acid methyl ester, (R)-3-[(tert-Butyldimethylsilanyl)oxy]-5-oxo-6-(triphenylphosphanylidene)hexanoic acid methyl ester, R-(-)-Oxo-3-(tert-butyldimethylsilyloxy)-6-triphenylphosphoranediylhexanoic acid methyl ester, Hexanoic acid, 3-[[(1,1-dimethylethyl)dimethylsilyl]oxy]-5-oxo-6-(triphenylphosphoranylidene)-, methyl ester, (3R)-, Methyl (3R)-3-[tert-butyl(dimethyl)silyl]oxy-5-oxo-6-(triphenyl-$l^{5}-phosphanylidene)hexanoate
IUPAC navn methyl (3R)-3-[tert-butyl(dimethyl)silyl]oxy-5-oxo-6-(triphenyl-λ^{5}-phosphanylidene)hexanoate
SMIL COC(=O)C[C@@H](CC(=O)C=P(c1ccccc1)(c2ccccc2)c3ccccc3)O[Si](C)(C)C(C)(C)C
Tekniske data
Kemisk navn eller materiale Methyl (3R)-3-[tert-Butyl(dimethyl)silyl]oxy-5-oxo-6-(triphenylphosphanylidene)hexanoate
Mængde 100 g
InChI formel InChI=1S/C31H39O4PSi/c1-31(2,3)37(5,6)35-26(23-30(33)34-4)22-25(32)24-36(27-16-10-7-11-17-27,28-18-12-8-13-19-28)29-20-14-9-15-21-29/h7-21,24,26H,22-23H2,1-6H3/t26-/m1/s1
Formel vægt 534.2355