Få mere at vide
Methyl 3-Keto-7alpha,12alpha-dihydroxy-5beta-cholanoate, TRC
Artikelnummer. 30472671
Skift visning
Mængde:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30472671 100 mg
30470226 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30472671 Leverandør Toronto Research Chemicals Leverandørnr. TRCM315055100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 14772-99-7
Molekylær formel C25H40O5
Molekylvægt (g/mol) 420.58
Synonym 7,12-Dihydroxy-3-oxocholan-24-oic Acid Methyl Ester, 7α,12α-Dihydroxy-3-oxo-5β-cholan-24-oic Acid Methyl Ester, 7α,12α-Dihydroxy-3-oxo-5β-cholanic Acid Methyl Ester, Methyl 3-Oxoisodeoxycholate
IUPAC navn methyl (4R)-4-[(5R,7R,8R,9S,10S,12S,13R,14S,17R)-7,12-dihydroxy-10,13-dimethyl-3-oxo-1,2,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]pentanoate
SMIL O[C@H](C[C@@](CC1=O)([H])[C@@](CC1)(C)[C@@]2([H])C[C@@H]3O)[C@@]2([H])[C@@](CC[C@]4([H])[C@H](C)CCC(OC)=O)([H])[C@]34C
Tekniske data
Kemisk navn eller materiale Methyl 3-Keto-7alpha,12alpha-dihydroxy-5beta-cholanoate
Mængde 100 mg
InChI formel InChI=1S/C25H40O5/c1-14(5-8-22(29)30-4)17-6-7-18-23-19(13-21(28)25(17,18)3)24(2)10-9-16(26)11-15(24)12-20(23)27/h14-15,17-21,23,27-28H,5-13H2,1-4H3/t14-,15+,17-,18+,19+,20-,21+,23+,24+,25-/m1/s1
Formel vægt 420.2876