Få mere at vide
Methyl-1-naphthalenemethylamine, TRC
Artikelnummer. 30136973
Skift visning
Mængde:
500 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136973 5 g
30134012 1 g
30010712 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30136973 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3231405G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 14489-75-9
Molekylær formel C12 H13 N
Molekylvægt (g/mol) 171.24
IUPAC navn N-methyl-1-naphthalen-1-ylmethanamine
SMIL CNCc1cccc2ccccc12
Tekniske data
Kemisk navn eller materiale Methyl-1-naphthalenemethylamine
Mængde 5 g
InChI formel InChI=1S/C12H13N/c1-13-9-11-7-4-6-10-5-2-3-8-12(10)11/h2-8,13H,9H2,1H3
Formel vægt 171.1048