Få mere at vide
Methoxy(4-methoxyphenyl)dimethylsilane, TRC
Artikelnummer. 30179040
Skift visning
Mængde:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30179040 100 mg
30179169 1 g
30179482 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30179040 Leverandør Toronto Research Chemicals Leverandørnr. TRCM269628100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 62244-48-8
Molekylær formel C10H16O2Si
Molekylvægt (g/mol) 196.32
Synonym Methoxy(p-methoxyphenyl)dimethylsilane
IUPAC navn methoxy-(4-methoxyphenyl)-dimethylsilane
SMIL COc1ccc(cc1)[Si](C)(C)OC
Tekniske data
Kemisk navn eller materiale Methoxy(4-methoxyphenyl)dimethylsilane
Mængde 100 mg
InChI formel InChI=1S/C10H16O2Si/c1-11-9-5-7-10(8-6-9)13(3,4)12-2/h5-8H,1-4H3
Formel vægt 196.092