Få mere at vide
Methanol, Supelco™
Gradient grade for liquid chromatography LiChrosolv™ Reag. Ph Eur, Reag. ChP
Brand:  Supelco 1060079030
 Se flere versioner af dette produkt
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Produktkode 15824730
  • 8000.00 DKK / 30 L
 Begrænset vare
Forventet forsendelse: 05-01-2027
Tilføj til indkøbskurv
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
LiChrosolv™ solvents were specially formulated to meet the demanding needs of analytical HPLC applications. With exceptional UV transmittance, minimal particle count, and controlled acidity and alkalinity, our solvents ensure reproducible separations every time. The low evaporation residue level further enhances their performance, making them the ideal choice for high-quality results. The product line includes both gradient grade and isocratic grade solvents, specifically designed to reduce the gradient effect during separations. This tailored approach allows for optimal consistency and accuracy in your analytical processes, helping you achieve the best possible outcomes in your chromatography work. Now Available with Data Matrix Code This high-quality HPLC-grade solvent now features a unique Data Matrix Code on its label, providing digital access to essential product information such as batch number, product number, and expiry date. Unlike standard barcodes, our DMCs feature a unique serial number for each bottle, enabling you to monitor and trace every bottle individually. With standard barcodes, all bottles from the same batch look identical. Our solution gives you true bottle-level traceability for greater control and compliance in the lab. Know more about our data matrix code-labeled HPLC-grade solvents.
TRUSTED_SUSTAINABILITY
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater