missing translation for 'onlineSavingsMsg'
Få mere at vide

meso-Lactide, TRC

Product Code. 30473808
Change view
Click to view available options
Mængde:
2.5 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30473808 2.5 mg
30466350 10 mg
30466349 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30473808 Supplier Toronto Research Chemicals Supplier No. TRCL1135952.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13076-19-2
Molekylær formel C6H8O4
Molekylvægt (g/mol) 144.13
Synonym (3R,6S)-3,6-Dimethyl-1,4-dioxane-2,5-dione, trans-3,6-Dimethyl-1,4-dioxane-2,5-dione, trans-3,6-Dimethyl-p-Dioxane-2,5-dione, trans-Lactide
SMIL O=C([C@@H](C)O1)O[C@@H](C)C1=O

Specifications

Kemisk navn eller materiale meso-Lactide
Renhedsgrad noter GC
Mængde 2.5 mg
InChI formel InChI=1S/C6H8O4/c1-3-5(7)10-4(2)6(8)9-3/h3-4H,1-2H3/t3-,4+
Formel vægt 144.0423
Procent renhed >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.