Få mere at vide
Meclizine Dihydrochloride Monohydrate, TRC
Artikelnummer. 30483581
Skift visning
Mængde:
5 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30483581 5 g
30469670 10 g
30470396 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30483581 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2027005G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 31884-77-2
Molekylær formel C25 H27 Cl N2 . 2 Cl H . H2 O
Molekylvægt (g/mol) 481.89
Synonym Piperazine, 1-[(4-chlorophenyl)phenylmethyl]-4-[(3-methylphenyl)methyl]-, hydrochloride, hydrate (1:2:1), Piperazine, 1-(p-chloro-α-phenylbenzyl)-4-(m-methylbenzyl)-, dihydrochloride, monohydrate (8CI), Piperazine, 1-[(4-chlorophenyl)phenylmethyl]-4-[(3-methylphenyl)methyl]-, dihydrochloride, monohydrate (9CI), Meclizine dihydrochloride monohydrate
IUPAC navn 1-[(4-chlorophenyl)-phenylmethyl]-4-[(3-methylphenyl)methyl]piperazine;hydrate;dihydrochloride
SMIL O.Cl.Cl.Cc1cccc(CN2CCN(CC2)C(c3ccccc3)c4ccc(Cl)cc4)c1
Tekniske data
Kemisk navn eller materiale Meclizine Dihydrochloride
Mængde 5 g
InChI formel InChI=1S/C25H27ClN2.2ClH.H2O/c1-20-6-5-7-21(18-20)19-27-14-16-28(17-15-27)25(22-8-3-2-4-9-22)23-10-12-24(26)13-11-23;;;/h2-13,18,25H,14-17,19H2,1H3;2*1H;1H2
Formel vægt 480.1502