Learn More
Hirschmann™ Measuring flask Class A with pre-packing regulation
Achieve precise standard solutions. Hirschmann™ Measuring flask Class A with pre-packing regulation is standardized to contain or to deliver a definite amount of liquid. Flask has blue graduations.
Brand:  Hirschmann™ 2950170
Additional Details : Weight : 0.25000kg
 View more versions of this product
Product Code. 11833212
  • 1130.00 DKK / Pack of 2
Estimated Shipment: 05-06-2024
to see stock.
Add to basket
Description
Description
  • Class A
  • Pursuant to the rules of the pre-packing regulation
  • Certified conformity
  • Blue graduation
  • Non-ground, rimmed
  • Printed scale on the neck for simple and rapid identification of deviation of actual value from normal value
  • Calibrated for IN
Specifications
Specifications
25 mL
0.5 mL
Yes
23 to 27 mL
110 mm
A