missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
ME2 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67457-1-IG-150UL
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
The ME2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ME2. This antibody recognizes human, mouse, rat antigen. The ME2 antibody has been validated for the following applications: WB, IHC, ELISA analysis.Tekniske data
| ME2 | |
| Monoclonal | |
| 900 μg/mL | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 4200 | |
| Store at -20°C. | |
| Monoclonal Antibody | |
| IgG2a |
| Western Blot, Immunohistochemistry, ELISA | |
| 6E6E2 | |
| Unconjugated | |
| Recombinant Protein | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| 54 kDa, 65 kDa | |
| Liquid |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion